4-Cyano-3-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Cyano-3-methoxyphenyl)boronic acid

A241849-1G A241849-250MG A241849-5G
76815-424EA 129.43 USD
76815-424 76815-426 76815-428
4-Cyano-3-methoxyphenylboronic acid 97%
4-Cyano-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
MW: 176.965 g/mol
MDL Number: MFCD10696655
CAS Number: 677777-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR