N-(2-(2,4,6-Trichlorophenoxy)ethyl)propan-1-amine 98%

Supplier: Ambeed
Warning

A241004-100G A241004-1G A241004-25G A241004-5G
76815-408EA 435 USD
76815-408 76815-410 76815-412 76815-414 76815-416 76815-418
N-(2-(2,4,6-Trichlorophenoxy)ethyl)propan-1-amine 98%
N-(2-(2,4,6-Trichlorophenoxy)ethyl)propan-1-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₄Cl₃NO
MDL Number: MFCD09751307
CAS Number: 67747-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR