4-Chloro-2-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-α,α,α-trifluoro-o-toluic acid

A240897-100G A240897-10G A240897-1G A240897-250MG A240897-5G A240897-25G
76815-398EA 442.4 USD
76815-398 76815-400 76815-402 76815-404 76815-406 77672-148
4-Chloro-2-(trifluoromethyl)benzoic acid 97%
4-Chloro-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
MDL Number: MFCD03094162
CAS Number: 142994-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR