Ethyl-trans-(ethoxymethylene)acetate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl trans-3-ethoxyacrylate, Ethyl (E)-3-Ethoxyacrylate

A239614-100G A239614-1G A239614-25G A239614-5G
76815-362EA 119 USD
76815-362 76815-364 76815-366 76815-368
Ethyl-trans-(ethoxymethylene)acetate 96%
Ethyl-trans-(ethoxymethylene)acetate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MDL Number: MFCD00009863
CAS Number: 5941-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR