1-Methyl-2-nitro-1H-imidazole 97%

Supplier: Ambeed
Warning

A239032-100MG A239032-1G A239032-250MG A239032-5G
76815-346EA 17 USD
76815-346 76815-348 76815-350 77883-634
1-Methyl-2-nitro-1H-imidazole 97%
1-Methyl-2-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
MDL Number: MFCD01721434
CAS Number: 1671-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR