3-Iodo-6-methyl-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A238501-100MG A238501-250MG A238501-1G
76815-338EA 89 USD
76815-338 76815-340 77672-134
3-Iodo-6-methyl-1H-pyrrolo[2,3-b]pyridine 97%
3-Iodo-6-methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇IN₂
MDL Number: MFCD09880117
CAS Number: 1000340-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR