Ethyl 5-chloro-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A238317-100MG A238317-1G A238317-250MG A238317-5G
76815-332EA 28.65 USD
76815-332 76815-334 76815-336 77612-314
Ethyl 5-chloro-1H-pyrazole-4-carboxylate 97%
Ethyl 5-chloro-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O₂
MW: 174.58 g/mol
MDL Number: MFCD08689931
CAS Number: 1393667-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR