Methyl 5-bromo-2-hydroxypyrimidine-4-carboxylate 97%

Supplier: Ambeed
Warning

A238272-100MG A238272-250MG
76815-328EA 82.76 USD
76815-328 76815-330
Methyl 5-bromo-2-hydroxypyrimidine-4-carboxylate 97%
Methyl 5-bromo-2-hydroxypyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₃
MW: 233.02 g/mol
MDL Number: MFCD09754091
CAS Number: 71933-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR