Ethyl 3-chloro-6-methylpyridazine-4-carboxylate 95%

Supplier: Ambeed
Warning

A237799-100MG A237799-250MG
76815-314EA 56 USD
76815-314 76815-316
Ethyl 3-chloro-6-methylpyridazine-4-carboxylate 95%
Ethyl 3-chloro-6-methylpyridazine-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MDL Number: MFCD26523210
CAS Number: 1445-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR