1H-Pyrrolo[2,3-b]pyridine-3-propanoic acid 95%

Supplier: Ambeed
Warning

A237062-1G A237062-250MG
76815-302EA 147 USD
76815-302 76815-304
1H-Pyrrolo[2,3-b]pyridine-3-propanoic acid 95%
1H-Pyrrolo[2,3-b]pyridine-3-propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD09756478
CAS Number: 27663-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR