3-Bromo-5-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A236504-100MG A236504-10G A236504-1G A236504-250MG A236504-25G A236504-5G A236504-100G
76815-282EA 5 USD
76815-282 76815-284 76815-286 76815-288 76815-290 76815-292 77672-106
3-Bromo-5-methoxybenzaldehyde 98%
3-Bromo-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MDL Number: MFCD06797975
CAS Number: 262450-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR