2,5-Dichloro-3-methylpyridine 98%

Supplier: Ambeed
Warning

A234661-100G A234661-10G A234661-25G A234661-5G A234661-1G
76815-254EA 458.31 USD
76815-254 76815-256 76815-258 76815-260 77672-094
2,5-Dichloro-3-methylpyridine 98%
2,5-Dichloro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N
MW: 162.0166 g/mol
MDL Number: MFCD03840754
CAS Number: 59782-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR