(E)-Methyl 3-(pyridin-4-yl)acrylate 97%

Supplier: Ambeed
Warning

A234139-1G A234139-250MG A234139-5G
76815-242EA 48 USD
76815-242 76815-244 77672-084
(E)-Methyl 3-(pyridin-4-yl)acrylate 97%
(E)-Methyl 3-(pyridin-4-yl)acrylate

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MDL Number: MFCD00023636
CAS Number: 81124-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR