Penta-O-acetyl-β-D-glucopyranose 97%

Supplier: Ambeed
Warning

Synonyms: β-D-Glucose Pentaacetate, Pentaacetyl-β-D-glucose, B-D-Glucose Pentaacetate, (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

A233926-100G A233926-25G A233926-500G A233926-1000G
76815-236EA 28 USD
76815-236 76815-238 76815-240 77672-082
Penta-O-acetyl-β-D-glucopyranose 97%
Penta-O-acetyl-beta-D-glucopyranose

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₁₁
MDL Number: MFCD00006597
CAS Number: 604-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR