3-Hydroxy-5-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A233392-100MG A233392-250MG A233392-5G A233392-1G
76815-226EA 39 USD
76815-226 76815-228 76815-230 77672-072
3-Hydroxy-5-methylbenzaldehyde 98%
3-Hydroxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.148 g/mol
MDL Number: MFCD16999707
CAS Number: 60549-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR