(E)-3-(p-Tolyl)acrylic acid 99%

Supplier: Ambeed
Warning

A233318-100G A233318-10G A233318-25G A233318-500G A233318-5G
76815-216EA 65 USD
76815-216 76815-218 76815-220 76815-222 76815-224
(E)-3-(p-Tolyl)acrylic acid 99%
(E)-3-(p-Tolyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD00002697
CAS Number: 940-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR