2-(2-Methoxyphenyl)ethanol 95%

Supplier: Ambeed
Warning

A233314-1G A233314-250MG A233314-25G A233314-5G A233314-10G
76815-208EA 15 USD
76815-208 76815-210 76815-212 76815-214 77883-614
2-(2-Methoxyphenyl)ethanol 95%
2-(2-Methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00004512
CAS Number: 7417-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR