4-Carbamoyl-2-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Carbamoyl-2-fluorophenyl)boronic acid

A233065-1G A233065-250MG A233065-5G
76815-202EA 160.2 USD
76815-202 76815-204 76815-206
4-Carbamoyl-2-fluorophenylboronic acid 98%
4-Carbamoyl-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BFNO₃
MW: 182.9500 g/mol
MDL Number: MFCD09800867
CAS Number: 874289-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR