tert-Butyl-N-(1,3-thiazol-2-yl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl thiazol-2-ylcarbamate

A232629-1G A232629-250MG A232629-25G A232629-5G A232629-100G
76815-190EA 6 USD
76815-190 76815-192 76815-194 76815-196 77883-611
tert-Butyl-N-(1,3-thiazol-2-yl)carbamate 97%
tert-Butyl-N-(1,3-thiazol-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD09038327
CAS Number: 170961-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR