1-Ethyl-3-methylimidazolium chloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride

A232266-100G A232266-25G A232266-5G A232266-500G A232266-1000G
76815-184EA 59.42 USD
76815-184 76815-186 76815-188 77568-252 77672-068
1-Ethyl-3-methylimidazolium chloride 97%
1-Ethyl-3-methylimidazolium chloride

Precaución: For research use only.

Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
MDL Number: MFCD00074843
CAS Number: 65039-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR