1,3-Bis(3-(dimethylamino)propyl)urea 97%

Supplier: Ambeed
Warning

A231860-100G A231860-10G A231860-25G A231860-500G
76815-166EA 84.88 USD
76815-166 76815-168 76815-170 76815-172
1,3-Bis(3-(dimethylamino)propyl)urea 97%
1,3-Bis(3-(dimethylamino)propyl)urea

Precaución: for Research Use Only

Formula: C₁₁H₂₆N₄O
MW: 230.35 g/mol
MDL Number: MFCD18970150
CAS Number: 52338-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR