Methyl 6-bromo-3-fluoropicolinate 97%

Supplier: Ambeed
Warning

A231574-100MG A231574-5G A231574-1G
76815-152EA 48 USD
76815-152 77672-056 77883-606
Methyl 6-bromo-3-fluoropicolinate 97%
Methyl 6-bromo-3-fluoropicolinate

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD14698123
CAS Number: 1214332-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR