Methyl 3-amino-6-bromopicolinate 97%

Supplier: Ambeed
Warning

A230295-100MG A230295-1G
76815-134EA 22 USD
76815-134 77883-602
Methyl 3-amino-6-bromopicolinate 97%
Methyl 3-amino-6-bromopicolinate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MDL Number: MFCD11656228
CAS Number: 866775-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR