(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-oxo-4-(tritylamino)butanoic acid 95%

Supplier: Ambeed
Warning

A230162-250MG A230162-5G A230162-1G A230162-100MG
76815-126EA 48 USD
76815-126 77550-490 77612-238 77672-042
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-oxo-4-(tritylamino)butanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-oxo-4-(tritylamino)butanoic acid

Precaución: for Research Use Only

Formula: C₃₉H₃₄N₂O₅
MDL Number: MFCD22200773
CAS Number: 941296-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR