7,10-Dimethoxy-10-DAB III 98%

Supplier: Ambeed
Warning

A229690-1G A229690-250MG A229690-5G A229690-100MG
76815-120EA 144 USD
76815-120 76815-122 76815-124 77883-601
7,10-Dimethoxy-10-DAB III 98%
7,10-Dimethoxy-10-DAB III

Precaución: for Research Use Only

Formula: C₃₁H₄₀O₁₀
MDL Number: MFCD23100710
CAS Number: 183133-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR