3-(Ethylcarbamoyl)-4-fluorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-(Ethylcarbamoyl)-4-fluorophenyl)boronic acid, 3-(Ethylcarbamoyl)-4-fluorophenylboronic acid

A228755-1G A228755-250MG A228755-5G
76815-106EA 103.97 USD
76815-106 76815-108 76815-110
3-(Ethylcarbamoyl)-4-fluorobenzeneboronic acid 98%
3-(Ethylcarbamoyl)-4-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BFNO₃
MW: 210.998 g/mol
MDL Number: MFCD08235039
CAS Number: 874219-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR