(2-Methyl-1H-pyrrolo[2,3-b]pyridin-4-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A227992-100MG A227992-250MG
76815-092EA 46 USD
76815-092 77672-012
(2-Methyl-1H-pyrrolo[2,3-b]pyridin-4-yl)boronic acid ≥95%
(2-Methyl-1H-pyrrolo[2,3-b]pyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MDL Number: MFCD16036274
CAS Number: 1014614-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR