4-(Prop-2-yn-1-yl)morpholine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(2-Propyn-1-yl)morpholine, 4-Propargylmorpholine
A227243-100G
A227243-10G
A227243-1G
A227243-250MG
A227243-5G
76815-062EA
324.64
USD
76815-062
76815-064
76815-066
76815-068
76815-070
4-(Prop-2-yn-1-yl)morpholine 97%
4-(Prop-2-yn-1-yl)morpholine
Precaución: for Research Use Only
Formula:
C₇H₁₁NO MW: 125.17 g/mol |
MDL Number:
MFCD00510919 CAS Number: 5799-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...