2-Amino-4-methylpentan-1-ol 97%

Supplier: Ambeed
Warning

A227059-10G A227059-25G A227059-5G A227059-1G
76815-056EA 103.97 USD
76815-056 76815-058 76815-060 77671-996
2-Amino-4-methylpentan-1-ol 97%
2-Amino-4-methylpentan-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
MW: 117.18 g/mol
MDL Number: MFCD00066926
CAS Number: 502-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR