(R)-2-Cyclohexyl-2-hydroxyacetic acid 95%

Supplier: Ambeed
Warning

A226240-100MG A226240-1G A226240-250MG A226240-5G
76815-044EA 29 USD
76815-044 76815-046 76815-048 77883-588
(R)-2-Cyclohexyl-2-hydroxyacetic acid 95%
(R)-2-Cyclohexyl-2-hydroxyacetic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MDL Number: MFCD00009824
CAS Number: 53585-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR