4-Fluoro-1H-indazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A225936-100MG A225936-1G A225936-250MG
76815-036EA 44.56 USD
76815-036 76815-038 76815-040
4-Fluoro-1H-indazole-5-carboxylic acid 97%
4-Fluoro-1H-indazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₂
MDL Number: MFCD11007930
CAS Number: 1041481-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR