8-Oxa-3-aza-bicyclo[3.2.1]octane 98%

Supplier: Ambeed
Danger

Synonyms: 8-Oxa-3-azabicyclo[3.2.1]octane

A225534-100MG A225534-250MG A225534-1G
76815-030EA 63.66 USD
76815-030 76815-032 77550-464
8-Oxa-3-aza-bicyclo[3.2.1]octane 98%
8-Oxa-3-aza-bicyclo[3.2.1]octane

Precaución: for Research Use Only

Formula: C₆H₁₁NO
MW: 113.158 g/mol
MDL Number: MFCD09836264
CAS Number: 280-13-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR