Methyl 4-iodo-1-methyl-1H-pyrazole-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A225413-100MG
A225413-250MG
A225413-5G
A225413-1G
76815-020EA
11
USD
76815-020
76815-022
76815-024
77671-990
Methyl 4-iodo-1-methyl-1H-pyrazole-3-carboxylate 97%
Methyl 4-iodo-1-methyl-1H-pyrazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₇IN₂O₂ |
MDL Number:
MFCD19703827 CAS Number: 75092-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...