1-(6-Fluoropyridin-2-yl)ethanone 98%

Supplier: Ambeed
Warning

A225091-100MG A225091-1G A225091-250MG
76815-012EA 24 USD
76815-012 76815-014 76815-016
1-(6-Fluoropyridin-2-yl)ethanone 98%
1-(6-Fluoropyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₆FNO
MDL Number: MFCD09263901
CAS Number: 501009-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR