4,6-Dichloro-1-methyl-1H-imidazo[4,5-c]pyridine 97%

Supplier: Ambeed
Warning

A224754-100MG A224754-250MG A224754-1G
76814-998EA 18 USD
76814-998 76815-000 77612-182
4,6-Dichloro-1-methyl-1H-imidazo[4,5-c]pyridine 97%
4,6-Dichloro-1-methyl-1H-imidazo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅Cl₂N₃
MDL Number: MFCD21603853
CAS Number: 887147-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR