5,6-Dichloropyridin-2-amine 98%

Supplier: Ambeed
Warning

A224289-250MG A224289-25G A224289-5G A224289-1G
76814-986EA 26.53 USD
76814-986 76814-988 76814-990 77593-460
5,6-Dichloropyridin-2-amine 98%
5,6-Dichloropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MW: 163.005 g/mol
MDL Number: MFCD17012012
CAS Number: 1192814-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR