Thieno[3,2-b]thiophene-2-carbonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-Cyanothieno[3,2-b]thiophene

A224065-100MG A224065-1G A224065-250MG A224065-5G
76814-974EA 27.59 USD
76814-974 76814-976 76814-978 76814-980
Thieno[3,2-b]thiophene-2-carbonitrile 95%
Thieno[3,2-b]thiophene-2-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₃NS₂
MW: 165.23 g/mol
MDL Number: MFCD08059461
CAS Number: 40985-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR