Methyl 3-formyl-1-methyl-1h-pyrazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-formyl-1-methyl-1H-pyrazole-5-carboxylate

A223215-100MG A223215-1G A223215-250MG
76814-942EA 28 USD
76814-942 76814-944 76814-946
Methyl 3-formyl-1-methyl-1h-pyrazole-5-carboxylate 97%
Methyl 3-formyl-1-methyl-1h-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD22376784
CAS Number: 1031351-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR