4-Bromo-5-fluoro-2-methoxyaniline 95%

Supplier: Ambeed
Warning

A222374-100G A222374-10G A222374-1G A222374-25G
76814-914EA 529.39 USD
76814-914 76814-916 76814-918 76814-920
4-Bromo-5-fluoro-2-methoxyaniline 95%
4-Bromo-5-fluoro-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFNO
MDL Number: MFCD21603958
CAS Number: 330794-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR