Methyl-3-chloro-4-iodobenzoate 95+%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-chloro-4-iodobenzoate

A221055-1G A221055-250MG A221055-5G
76814-884EA 98.67 USD
76814-884 76814-886 76814-888
Methyl-3-chloro-4-iodobenzoate 95+%
Methyl 3-chloro-4-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClIO₂
MW: 296.4895 g/mol
MDL Number: MFCD09800724
CAS Number: 874569-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR