6-Bromo-2-methylpyridazin-3(2H)-one 95%

Supplier: Ambeed
Warning

A220382-1G A220382-250MG A220382-5G
76814-858EA 17 USD
76814-858 76814-860 76814-862
6-Bromo-2-methylpyridazin-3(2H)-one 95%
6-Bromo-2-methylpyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MDL Number: MFCD12198437
CAS Number: 1123169-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR