cis-4-Carbomethoxy-cyclohexane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: cis-4-(Methoxycarbonyl)cyclohexanecarboxylic acid

A219729-100G A219729-10G A219729-1G A219729-250MG A219729-25G A219729-5G
76814-834EA 2120.74 USD
76814-834 76814-836 76814-838 76814-840 76814-842 76814-844
cis-4-Carbomethoxy-cyclohexane-1-carboxylic acid 95%
cis-4-Carbomethoxy-cyclohexane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
MW: 186.205 g/mol
MDL Number: MFCD01311246
CAS Number: 1011-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR