4,6-Dichloro-1-ethyl-1H-pyrazolo[3,4-d]pyrimidine 95%

Supplier: Ambeed
Warning

A216971-100MG A216971-1G A216971-250MG A216971-5G
76814-796EA 38.2 USD
76814-796 76814-798 76814-800 76814-802
4,6-Dichloro-1-ethyl-1H-pyrazolo[3,4-d]pyrimidine 95%
4,6-Dichloro-1-ethyl-1H-pyrazolo[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₄
MW: 217.05 g/mol
MDL Number: MFCD16249852
CAS Number: 864292-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR