4-Amino-2-chloro-3-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-methylpyridin-4-amine

A216957-1G A216957-250MG A216957-5G
76814-790EA 129.43 USD
76814-790 76814-792 76814-794
4-Amino-2-chloro-3-methylpyridine 97%
4-Amino-2-chloro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.586 g/mol
MDL Number: MFCD18415428
CAS Number: 79055-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR