9-(Bromomethyl)anthracene 98%

Supplier: Ambeed
Danger

A216919-100G A216919-100MG A216919-1G A216919-250MG A216919-25G A216919-5G
76814-780EA 1094.85 USD
76814-780 76814-782 76814-784 76814-786 76814-788 77550-402
9-(Bromomethyl)anthracene 98%
9-(Bromomethyl)anthracene

Precaución: for Research Use Only

Formula: C₁₅H₁₁Br
MW: 271.152 g/mol
MDL Number: MFCD00096212
CAS Number: 2417-77-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR