5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-hydroxy-1H-pyrazole-3-carboxylate

A216637-1G A216637-250MG A216637-25G A216637-5G A216637-10G
76814-764EA 18.04 USD
76814-764 76814-766 76814-768 77568-096 77671-938
5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester 97%
5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester

Precaución: For research use only.

Formula: C₆H₈N₂O₃
MW: 156.139 g/mol
MDL Number: MFCD09263987
CAS Number: 85230-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR