3-Methyl-2-pyridone 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-2-pyridinone, 2-Hydroxy-3-methylpyridine, 3-Methylpyridin-2(1H)-one

A216532-100G A216532-10G A216532-1G A216532-25G A216532-5G
76814-754EA 196.27 USD
76814-754 76814-756 76814-758 76814-760 76814-762
3-Methyl-2-pyridone 98%
3-Methyl-2-pyridone

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00039704
CAS Number: 1003-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR