3-Bromoquinolin-2-amine 97%

Supplier: Ambeed
Warning

A216222-100MG A216222-1G A216222-250MG
76814-748EA 18 USD
76814-748 76814-750 76814-752
3-Bromoquinolin-2-amine 97%
3-Bromoquinolin-2-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD18255384
CAS Number: 36825-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR