tert-Butyl-3-formyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-formyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

A216004-1G A216004-5G
76814-744EA 51.99 USD
76814-744 76814-746
tert-Butyl-3-formyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 97%
tert-Butyl-3-formyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₃
MW: 246.27 g/mol
MDL Number: MFCD11045434
CAS Number: 144657-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR