N-Carbobenzoxy-D-glutamic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-D-glutamic acid, N-Cbz-D-glutamic Acid, Z-D-Glu-OH, Cbz-D-Glu-OH

A215970-1G A215970-25G A215970-5G A215970-100G
76814-738EA 5 USD
76814-738 76814-740 76814-742 77612-118
N-Carbobenzoxy-D-glutamic acid 98%
N-Carbobenzoxy-D-glutamic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₆
MDL Number: MFCD00063193
CAS Number: 63648-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR